Europium(III) chloride hexahydrate 99.99% (trace metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Europium(III) chloride hydrate, Europium trichloride hexahydrate

AC19301-0010 AC19301-0050 AC19301-0250
TS19301-0010EA 56 USD
TS19301-0010 TS19301-0050 TS19301-0250
Europium(III) chloride hexahydrate 99.99% (trace metals basis)
Europium(III) chloride hexahydrate
Formula: Cl₃Eu·6H₂O
MW: 366.41
Melting Pt: 158
Density: 4.89 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149701
CAS Number: 13759-92-7
EINECS: 233-040-4

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Titration Complexometric 39.0 to 42.5 % (Eu)
Total conc. of imp.as REO ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR