Europium(III) chloride hexahydrate 99.9% (trace metals basis) -10 mesh
Supplier: Thermo Scientific Chemicals
Synonyms:
Europium(III) chloride hydrate, Europium trichloride hexahydrate
AC19960-0050
AC19960-0250
TS19960-0050EA
137
USD
TS19960-0050
TS19960-0250
Europium(III) chloride hexahydrate 99.9% (trace metals basis) -10 mesh
Europium(III) chloride hexahydrate
Formula:
Cl₃Eu·6H₂O MW: 366.41 g/mol Melting Pt: 850 °C Density: 4.89 g/cm³ |
MDL Number:
MFCD00149701 CAS Number: 13759-92-7 EINECS: 233-040-4 |
Specification Test Results
Appearance | White Crystals |
Total conc. of imp.as REO | ≤0.1 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...