tetra-Chloroauric(III) acid hydrate 99.999% (trace metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Gold(III) chloride hydrate, Hydrogen tetrachloroaurate(III) xhydrate, Hydrogen tetrachloroaurate (III) hydrate, Hydrogen tetrachloroaurate hydrate, Chloroauric acid, tetra-Chloroauric acid hydrate
AC43717-0010
AC43717-0050
TS43717-0010EA
267
USD
TS43717-0010
TS43717-0050
tetra-Chloroauric(III) acid hydrate 99.999% (trace metals basis)
tetra-Chloroauric(III) acid hydrate
Formula:
HAuCl₄·H₂O MW: 357.8 g/mol Melting Pt: 30 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00149903 CAS Number: 27988-77-8 UN: 3260 ADR: 8,II |
Specification Test Results
Appearance | Yellow to orange Crystals |
Assay | 49 to 51 % (Au) |
Total trace metal impurities | ≤10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...