Tin(IV) iodide 99+% -10 mesh

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tin tetraiodide

AC38856-0100 AC38856-0500
TS38856-0100EA 202 USD
TS38856-0100 TS38856-0500
Tin(IV) iodide 99+% -10 mesh
Tin(IV) iodide
Formula: I₄Sn
MW: 626.33 g/mol
Melting Pt: 144 °C
Storage Temperature: Ambient
MDL Number: MFCD00036272
CAS Number: 7790-47-8
UN: 3260
ADR: 8,II

Order Now

Specification Test Results

Appearance Brown Powder
Titration Argentometric ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR