Zinc sulphate heptahydrate 99% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC42460-0010
AC42460-0030
AC42460-0100
AC42460-5000
TS42460-0010EA
147
USD
TS42460-0010
TS42460-0030
TS42460-0100
TS42460-5000
Zinc sulphate heptahydrate 99% ACS
Zinc sulphate heptahydrate
Formula:
ZnSO₄·7H₂O Melting Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149894 CAS Number: 7446-20-0 EINECS: 231-793-3 UN: 3077 ADR: 9,III Merck Index: 12,10293 |
Specification Test Results
Appearance | White Crystals and/or chunks |
Titration Complexometric | 99.0 to 103.0 % |
pH | 4.4 to 6.0 (5 % at 25°C) |
Chloride (Cl) | ≤5 ppm |
Nitrate (NO3) | ≤20 ppm |
Ammonium (NH4) | ≤10 ppm |
Calcium (Ca) | ≤50 ppm |
Iron (Fe) | ≤10 ppm |
Lead (Pb) | ≤30 ppm |
Magnesium (Mg) | ≤50 ppm |
Manganese (Mn) | ≤3 ppm |
Potassium (K) | ≤100 ppm |
Sodium (Na) | ≤500 ppm |
Insoluble matter | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...