Zinc iodide 99.999% (trace metals basis), Extra Pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC21276-0050
AC21276-0250
TS21276-0050EA
66
USD
TS21276-0050
TS21276-0250
Zinc iodide 99.999% (trace metals basis), Extra Pure
Zinc iodide
Formula:
I₂Zn Melting Pt: 446 °C Density: 4.74 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011299 CAS Number: 10139-47-6 EINECS: 233-396-0 UN: 3260 ADR: 8,II Merck Index: 12,10269 |
Specification Test Results
Appearance | White to light yellow Powder |
Total trace metal impurities | ≤0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...