Zinc fluoride 98%

Supplier: Thermo Scientific Chemicals
Danger

AC36391-0250 AC36391-2500
TS36391-0250EA 55 USD
TS36391-0250 TS36391-2500
Zinc fluoride 98%
Zinc fluoride
Formula: F₂Zn
MW: 103.39 g/mol
Boiling Pt: 1500 °C (1013 hPa)
Melting Pt: 872…910 °C
MDL Number: MFCD00011298
CAS Number: 7783-49-5
EINECS: 232-001-9

Order Now

Specification Test Results

Appearance White to off-white Powder
Fluoride (F) 36.0 to 37.8 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR