Trioctylsilane 95%

Supplier: Thermo Scientific Chemicals
Warning

AC43490-0050 AC43490-0250
TS43490-0050EA 64 USD
TS43490-0050 TS43490-0250
Trioctylsilane 95%
Trioctylsilane
Formula: C₂₄H₅₂Si
MW: 368.76 g/mol
Boiling Pt: 163…165 °C (.2 mmHg)
Density: 0.821 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00053812
CAS Number: 18765-09-8

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR