Titanium(IV) oxide anatase 98+%, powder
Supplier: Thermo Scientific Chemicals
AC21358-0010
AC21358-1000
TS21358-0010EA
126
USD
TS21358-0010
TS21358-1000
Titanium(IV) oxide anatase 98+%, powder
Titanium(IV) oxide anatase
Formula:
TiO₂ Melting Pt: 1855 °C |
MDL Number:
MFCD00011269 CAS Number: 1317-70-0 EINECS: 215-280-1 |
Specification Test Results
Appearance | White Powder |
Assay | ≥98.0 % |
Particle size | ≤0.05 % (residue on 44 µm sieve) |
pH | 7.3 to 8.3 (suspension in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...