Tetrapropylammonium bromide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: TPABr

AC20937-0025 AC20937-0250 AC20937-1000 AC20937-5000
TS20937-0025EA 821 USD
TS20937-0025 TS20937-0250 TS20937-1000 TS20937-5000
Tetrapropylammonium bromide 98%
Tetrapropylammonium bromide
Formula: C₁₂H₂₈BrN
MW: 266.27 g/mol
Melting Pt: 266…272 °C
Storage Temperature: Ambient
MDL Number: MFCD00011840
CAS Number: 1941-30-6
EINECS: 217-727-6

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals
Infrared spectrum Conforms
Titration Argentometric 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR