Tetramethylsilane ACS for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC44590-0250
AC44590-1000
TS44590-0250EA
55
USD
TS44590-0250
TS44590-1000
Tetramethylsilane ACS for NMR spectroscopy
Tetramethylsilane
Formula:
C₄H₁₂Si Boiling Pt: 26…28 °C (760 mmHg) Melting Pt: –99 °C Density: 0.64 g/cm³ Flash Pt: –27 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008274 CAS Number: 75-76-3 UN: 2749 ADR: 3,I |
Specification Test Results
Appearance | Clear colorless Liquid |
Residue after evaporation | ≤0.05 % |
Suitability | (for 1H-NMR position ref.) Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...