Tetramethylammonium hydroxide pentahydrate 97%

Supplier: Thermo Scientific Chemicals
Danger

AC20752-0250 AC20752-1000 AC20752-5000
TS20752-0250EA 68 USD
TS20752-0250 TS20752-1000 TS20752-5000
Tetramethylammonium hydroxide pentahydrate 97%
Tetramethylammonium hydroxide pentahydrate
Formula: C₄H₁₃NO·5H₂O
MW: 181.23 g/mol
Melting Pt: 62…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00149566
CAS Number: 10424-65-4
EINECS: 200-882-9
UN: 3423
ADR: 8,II

Order Now

Specification Test Results

Appearance White to off-white Hygroscopic powder or crystals
Infrared spectrum Conforms
Melting point 62 to 73 °C
Titration with HCl ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR