Tetramethylammonium bromide 98%

Supplier: Thermo Scientific Chemicals
Danger

AC18763-1000 AC18763-5000
TS18763-1000EA 58 USD
TS18763-1000 TS18763-5000
Tetramethylammonium bromide 98%
Tetramethylammonium bromide
Formula: C₄H₁₂BrN
Boiling Pt: ∼360 °C (1013 hPa)
Melting Pt: 230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011626
CAS Number: 64-20-0
EINECS: 200-581-2
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Titration Argentometric 97.5 to 102.5 %
Water ≤1 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR