Tetraethylammonium bromide 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: Tetrylammonium bromide

AC22071-0025 AC22071-0500 AC22071-2500 AC22071-5000
TS22071-0025EA 1253 USD
TS22071-0025 TS22071-0500 TS22071-2500 TS22071-5000
Tetraethylammonium bromide 99+%
Tetraethylammonium bromide
Formula: C₈H₂₀BrN
MW: 210.16 g/mol
Melting Pt: 285 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011825
CAS Number: 71-91-0
EINECS: 200-769-4
Merck Index: 13,09274

Order Now

Specification Test Results

Appearance White to cream Crystalline powder or crystals
Infrared spectrum Conforms
Titration Argentometric ≥99.0 %
Loss on drying ≤1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR