Tetrabutylammonium chloride xhydrate 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC43206-0050
AC43206-0250
AC43206-1000
TS43206-0050EA
33
USD
TS43206-0050
TS43206-0250
TS43206-1000
Tetrabutylammonium chloride xhydrate 96%
Tetrabutylammonium chloride xhydrate
Formula:
C₁₆H₃₆ClN·xH₂O Melting Pt: 37…39 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011635 CAS Number: 37451-68-6 |
Specification Test Results
Appearance | Off-white Low melting solid |
Infrared spectrum | Conforms |
Titration Argentometric | ≥95.0 % (On dried substance) |
Water | ≤10.0 % (K.F.) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...