Tetrabutylammonium chloride 95% tech. contains also bromide
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC17412-0050
AC17412-0250
AC17412-1000
AC17412-5000
TS17412-0050EA
34
USD
TS17412-0050
TS17412-0250
TS17412-1000
TS17412-5000
Tetrabutylammonium chloride 95% tech. contains also bromide
Tetrabutylammonium chloride
Formula:
C₁₆H₃₆ClN MW: 277.92 g/mol Melting Pt: 83…86 °C Flash Pt: 110 Storage Temperature: Ambient |
MDL Number:
MFCD00011635 CAS Number: 1112-67-0 |
Specification Test Results
Appearance | White to brown Crystalline solid |
Infrared spectrum | Conforms |
Titration Argentometric | ≥94 % (on dry substance) |
Water | ≤4 % |
Bromide (Br) | ≤5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...