Strontium hydroxide octahydrate 98%, pure

Supplier: Thermo Scientific Chemicals
Danger

AC27023-0010 AC27023-0050
TS27023-0010EA 77 USD
TS27023-0010 TS27023-0050
Strontium hydroxide octahydrate 98%, pure
Strontium hydroxide octahydrate
Formula: H₂O₂Sr·8H₂O
MW: 265.76 g/mol
Melting Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00150395
CAS Number: 1311-10-0
EINECS: 242-367-1
UN: 3262
ADR: 8,II

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration with HCl ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR