Silver(II) oxide

Supplier: Thermo Scientific Chemicals
Danger

AC20785-0100 AC20785-0500
TS20785-0100EA 71 USD
TS20785-0100 TS20785-0500
Silver(II) oxide
Silver(II) oxide
Formula: AgO
MW: 123.87 g/mol
Melting Pt: >100 °C (dec.)
MDL Number: MFCD00044285
CAS Number: 1301-96-8
EINECS: 215-098-2
UN: 3085
ADR: 5.1,II
Merck Index: 12,08666

Order Now

Specification Test Results

Appearance Gray to black Powder
Assay 85.5 to 88 % (AG)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR