Mercury(II) iodide, red 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Mercury diiodide, Mercuric iodide
AC20574-0025
AC20574-0050
AC20574-1000
AC20574-5000
TS20574-0025EA
2322
USD
TS20574-0025
TS20574-0050
TS20574-1000
TS20574-5000
Mercury(II) iodide, red 99+%
Mercury(II) iodide
Formula:
HgI₂ MW: 454.4 g/mol Boiling Pt: 354 Melting Pt: 259 Storage Temperature: Ambient |
MDL Number:
MFCD00011044 CAS Number: 7774-29-0 EINECS: 231-873-8 UN: 1638 ADR: 6.1 II Merck Index: 13,05905 |
Specification Test Results
Appearance | Red Crystalline powder |
Titration Iodometric | ≥99.0 % |
Solubility | (in Potassium iodide soln.) Passes test |
Mercurous Mercury (as Hg) | ≤0.1 % |
Soluble Hg salts (as Hg) | ≤0.05 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...