Mercury(II) chloride 99.5%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Mercuric chloride, Mercury Chloride, Mercury dichloride
AC20143-0010
AC20143-5000
TS20143-0010EA
337
USD
TS20143-0010
TS20143-0250
TS20143-1000
TS20143-5000
Mercury(II) chloride 99.5%
Mercury(II) chloride
Formula:
HgCl₂ MW: 271.5 g/mol Boiling Pt: 302 Melting Pt: 273 Storage Temperature: Ambient |
MDL Number:
MFCD00011041 CAS Number: 7487-94-7 EINECS: 231-299-8 UN: 1624 ADR: 6.1 II Merck Index: 13,05901 |
Specification Test Results
Appearance | White Crystalline powder or crystals (May contain black particles) |
Titration Complexometric | ≥99.4 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...