Titanium(IV) ethanolate 85% tech. contains 5-15% isopropanol
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC31254-0500
AC31254-2500
TS31254-0500EA
66
USD
TS31254-0500
TS31254-2500
Titanium(IV) ethanolate 85% tech. contains 5-15% isopropanol
Titanium(IV) ethanolate
Formula:
C₈H₂₀O₄Ti Boiling Pt: 150…152 °C (10 mmHg) Density: 1.08 g/cm³ Flash Pt: 29 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009071 CAS Number: 3087-36-3 EINECS: 221-410-8 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless to dark yellow Liquid |
Infrared spectrum | Conforms |
Assay | ≥85 % |
Isopropyl alcohol | 5 to 15 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...