Trimethyl borate 99%, AcroSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC34612-1000
AC34612-8000
TS34612-1000EA
34
USD
TS34612-1000
TS34612-8000
Trimethyl borate 99%, AcroSeal™
Trimethyl borate
Formula:
C₃H₉BO₃ Boiling Pt: 68…69 °C (760 mmHg) Melting Pt: –34 °C Density: 0.915 g/cm³ Flash Pt: –8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008346 CAS Number: 121-43-7 UN: 2416 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...