Trimethyl borate 99%

Supplier: Thermo Scientific Chemicals
Danger

AC14012-0025 AC14012-1000 AC14012-5000
TS14012-0025EA 213 USD
TS14012-0025 TS14012-1000 TS14012-5000
Trimethyl borate 99%
Trimethyl borate
Formula: C₃H₉BO₃
MW: 103.91 g/mol
Boiling Pt: 68…69 °C (760 mmHg)
Melting Pt: –34 °C
Density: 0.915 g/cm³
Flash Pt: –8 °C
Storage Temperature: Ambient
MDL Number: MFCD00008346
CAS Number: 121-43-7
EINECS: 204-468-9
UN: 2416
ADR: 3,II
Merck Index: 12,09842

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3562 to 1.3582 (20°C, 589 nm)
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR