Triethyl borate 97%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC42865-1000 AC42865-8000
TS42865-1000EA 52 USD
TS42865-1000 TS42865-8000
Triethyl borate 97%, AcroSeal™
Triethyl borate
Formula: C₆H₁₅BO₃
MW: 145.99 g/mol
Boiling Pt: 117 °C (1013 hPa)
Melting Pt: –84.5 °C
Density: 0.858 g/cm³
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00012846
CAS Number: 150-46-9
UN: 1176
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3705 to 1.3775 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR