Octyltriethoxysilane 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Triethoxy(octyl)silane, Triethoxyoctylsilane

AC33808-0250 AC33808-1000
TS33808-0250EA 22 USD
TS33808-0250 TS33808-1000
Octyltriethoxysilane 97%
Octyltriethoxysilane
Formula: C₁₄H₃₂O₃Si
MW: 276.49 g/mol
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: –40 °C
Density: 0.88 g/cm³
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00039883
CAS Number: 2943-75-1
EINECS: 220-941-2

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4160 to 1.4180 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR