Zinc stearate
Supplier: Thermo Scientific Chemicals
Synonyms:
Stearic acid zinc salt, Zinc(II) stearate
AC26378-0010
AC26378-0050
AC26378-0200
TS26378-0010EA
49
USD
TS26378-0010
TS26378-0050
TS26378-0200
Zinc stearate
Zinc stearate
Formula:
C₃₆H₇₀O₄Zn MW: 632.34 g/mol Melting Pt: 140 Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013031 CAS Number: 557-05-1 EINECS: 209-151-9 Merck Index: 13,10212 |
|
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
Titration Complexometric | 12.5 to 14.0 % (ZnO) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...