Nickel (II) oxide, black 76% Ni

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Nickel monoxide, Nickel oxide, Bunsenite

AC22312-0025 AC22312-0050 AC22312-1000 AC22312-5000
TS22312-0025EA 597 USD
TS22312-0025 TS22312-0050 TS22312-1000 TS22312-5000
Nickel (II) oxide, black 76% Ni
Nickel (II) oxide
Formula: NiO
Melting Pt: 1960 °C
Storage Temperature: Ambient
MDL Number: MFCD00011145
CAS Number: 1313-99-1
EINECS: 215-215-7
Merck Index: 13,06540

Order Now

Specification Test Results

Appearance Dark gray-black Powder
Assay 75 to 77 % (Ni)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR