Nickel(II) chloride hexahydrate 99.999% (trace metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC19357-0050
AC19357-0250
AC19357-1000
TS19357-0050EA
78
USD
TS19357-0050
TS19357-0250
TS19357-1000
Nickel(II) chloride hexahydrate 99.999% (trace metals basis)
Nickel(II) chloride hexahydrate
Formula:
NiCl₂·6H₂O Melting Pt: 140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149809 CAS Number: 7791-20-0 EINECS: 231-743-0 UN: 3288 ADR: 6.1,III Merck Index: 13,06529 |
Specification Test Results
Appearance | Green Crystalline powder or crystals and/or chunks |
Total trace metal impurities | ≤0.001 % |
Nickel (Ni) | 24 to 26 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...