Nickel(II) chloride, anhydrous 99.99% (trace metals basis), powder

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Nickel dichloride

AC37835-0050 AC37835-0250
TS37835-0050EA 154 USD
TS37835-0050 TS37835-0250
Nickel(II) chloride, anhydrous 99.99% (trace metals basis), powder
Nickel(II) chloride
Formula: Cl₂Ni
MW: 129.6 g/mol
Melting Pt: 1001 °C
Storage Temperature: Ambient
MDL Number: MFCD00011142
CAS Number: 7718-54-9
EINECS: 231-743-0
UN: 3288
ADR: 6.1,III
Merck Index: 12,06588

Order Now

Specification Test Results

Appearance Yellow to orange Powder
Titration Complexometric 44.6 to 46.0 % (Ni)
Total trace metal impurities ≤100 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR