Ethyl formate 98+%, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Formic acid ethyl ester

AC15067-0010 AC15067-0025 AC15067-0251 AC15067-5000
TS15067-0010EA 120 USD
TS15067-0010 TS15067-0025 TS15067-0251 TS15067-5000
Ethyl formate 98+%, pure
Ethyl formate
Formula: C₃H₆O₂
Boiling Pt: 54 °C (760 mmHg)
Melting Pt: –80 °C
Density: 0.917 g/cm³
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00003294
CAS Number: 109-94-4
EINECS: 203-721-0
UN: 1190
ADR: 3,II
Merck Index: 13,03841

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3580 to 1.3630 (20°C, 589 nm)
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR