Sodium methoxide 30% (w/w) 5.4 M in methanol, AcroSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC42836-1000
AC42836-8000
TS42836-1000EA
35
USD
TS42836-1000
TS42836-8000
Sodium methoxide 30% (w/w) 5.4 M in methanol, AcroSeal™
Sodium methoxide
Formula:
CH₃NaO Melting Pt: 1…5 °C Density: 0.97 g/cm³ Flash Pt: 33 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012179 CAS Number: 124-41-4 UN: 1289 ADR: 3,III |
Specification Test Results
Appearance | Colorless to light yellow Solution |
Strength | 5.3 to 5.6 M |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...