Manganese(IV) oxide 99% -10 mesh

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Manganese dioxide, Manganese(IV) oxide, activated, Manganese peroxide, Mangan(IV)-oxid

AC22258-0025 AC22258-1000 AC22258-5000
TS22258-0025EA 568 USD
TS22258-0025 TS22258-1000 TS22258-5000
Manganese(IV) oxide 99% -10 mesh
Manganese(IV) oxide
Formula: MnO₂
MW: 86.94 g/mol
Melting Pt: 535 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003463
CAS Number: 1313-13-9
EINECS: 215-202-6
Merck Index: 14,05730

Order Now

Specification Test Results

Appearance Gray to black Powder
Titration with Na2S2O3 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR