Manganese(IV) oxide 80-85%, powder
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Manganese dioxide, Manganese(IV) oxide, activated, Manganese peroxide, Mangan(IV)-oxid
AC21349-0010
AC21349-0025
AC21349-0250
TS21349-0010EA
116
USD
TS21349-0010
TS21349-0025
TS21349-0250
Manganese(IV) oxide 80-85%, powder
Manganese(IV) oxide
Formula:
MnO₂ MW: 86.94 g/mol Melting Pt: 535 (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00003463 CAS Number: 1313-13-9 EINECS: 215-202-6 Merck Index: 14,05730 |
Specification Test Results
Appearance | Dark gray Powder |
Loss on drying | ≤0.5 % (105°C) |
Titration with Na2S2O3 | ≥80 % |
Silicate (as SiO2) | ≤7 % |
Iron (Fe) | ≤1 % |
Insoluble matter | ≤10 % (in HCl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...