Magnesium bromide hexahydrate 99%, extra pure

Supplier: Thermo Scientific Chemicals
Warning

AC44625-0010 AC44625-2500
TS44625-0010EA 146 USD
TS44625-0010 TS44625-0050 TS44625-2500
Magnesium bromide hexahydrate 99%, extra pure
Magnesium bromide hexahydrate
Formula: Br₂Mg·6H₂O
MW: 292.2 g/mol
Melting Pt: 146 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149780
CAS Number: 13446-53-2

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Complexometric ≥98.5 %
pH 6 to 8 (5 % at 20°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR