Magnesium bromide hexahydrate 99+% for analysis

Supplier: Thermo Scientific Chemicals
Warning

AC20251-0010 AC20251-2500
TS20251-0010EA 174 USD
TS20251-0010 TS20251-2500
Magnesium bromide hexahydrate 99+% for analysis
Magnesium bromide hexahydrate
Formula: Br₂Mg·6H₂O
Melting Pt: 165 °C (dec.)
MDL Number: MFCD00149780
CAS Number: 13446-53-2
EINECS: 232-170-9

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Complexometric ≥99.0 %
pH 6 to 8 (5 % at 20°C)
Chloride (Cl) ≤500 ppm
Sulfate (SO4) ≤200 ppm
Cadmium (Cd) ≤50 ppm
Copper (Cu) ≤50 ppm
Iron (Fe) ≤50 ppm
Lead (Pb) ≤50 ppm
Zinc (Zn) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR