Copper(II) chloride, anhydrous 99%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Copper dichloride, Cupric chloride

AC20653-0010 AC20653-0025 AC20653-2500
TS20653-0010EA 208 USD
TS20653-0010 TS20653-0025 TS20653-2500
Copper(II) chloride, anhydrous 99%, Extra Pure
Copper(II) chloride
Formula: Cl₂Cu
MW: 134.45 g/mol
Boiling Pt: 993 °C (760 mmHg)
Melting Pt: 498 °C
Storage Temperature: Ambient
MDL Number: MFCD00010972
CAS Number: 7447-39-4
EINECS: 231-210-2
UN: 2802
ADR: 8,III
Merck Index: 13,02660

Order Now

Specification Test Results

Appearance Brown Crystalline powder
Assay ≥98.5 % (CuCl2)
Water ≤0.75 %
Impurity ≤0.75 % (Cupric nitrate)
Insoluble matter ≤0.25 % (in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR