Indium(III) bromide 99.99% (trace metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Indium tribromide
AC38903-0100
AC38903-0500
TS38903-0100EA
163
USD
TS38903-0100
TS38903-0500
Indium(III) bromide 99.99% (trace metals basis)
Indium(III) bromide
Formula:
Br₃In MW: 354.53 g/mol Boiling Pt: 656 °C (1013 hPa) Melting Pt: 220 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011057 CAS Number: 13465-09-3 EINECS: 236-692-8 |
Specification Test Results
Appearance | White to off-white Powder or crystals or granules and/or chunks |
Total trace metal impurities | ≤100 ppm |
Indium (In) | 31.7 to 33.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...