Holmium(III) oxide 99.9% (REO, rare earth oxide basis) -325 mesh
Supplier: Thermo Scientific Chemicals
Synonyms:
Diholmium trioxide, Holmium oxide
AC19714-0100
AC19714-0500
TS19714-0100EA
135
USD
TS19714-0100
TS19714-0500
Holmium(III) oxide 99.9% (REO, rare earth oxide basis) -325 mesh
Holmium(III) oxide
Formula:
Ho₂O₃ MW: 377.86 g/mol Boiling Pt: 2600 Melting Pt: 2400 Storage Temperature: Ambient |
MDL Number:
MFCD00011053 CAS Number: 12055-62-8 EINECS: 235-015-3 |
Specification Test Results
Appearance | Yellow to beige to light orange Powder |
Assay | ≥99.85 % (Ho2O3) TREO base |
Particle size | ≤325 mesh |
Loss on ignition | ≤1.0 % (1000°C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...