Germanium(IV) oxide 99.999% (trace metals basis), Electronic Grade

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Germanium dioxide

AC19000-0100 AC19000-0500 AC19000-2500
TS19000-0100EA 131 USD
TS19000-0100 TS19000-0500 TS19000-2500
Germanium(IV) oxide 99.999% (trace metals basis), Electronic Grade
Germanium(IV) oxide
Formula: GeO₂
MW: 104.61 g/mol
Melting Pt: 1115 °C
Storage Temperature: Ambient
MDL Number: MFCD00011030
CAS Number: 1310-53-8
EINECS: 215-180-8
Merck Index: 12,04417

Order Now

Specification Test Results

Appearance White Powder
Total trace metal impurities ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR