Gadolinium trichloride, anhydrous 99.9% (trace metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC38356-0050
AC38356-0250
TS38356-0050EA
180
USD
TS38356-0050
TS38356-0250
Gadolinium trichloride, anhydrous 99.9% (trace metals basis)
Gadolinium trichloride
Formula:
Cl₃Gd MW: 263.61 Melting Pt: 604…614 °C Density: 4.52 |
MDL Number:
MFCD00011024 CAS Number: 10138-52-0 EINECS: 233-386-6 |
Specification Test Results
Appearance | White to off-white Powder |
Titration Complexometric | 58.2 to 61.1 % (Gd) |
Total trace metal impurities | ≤1000 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...