Sodium acetate, anhydrous 99% for biochemistry
Supplier: Thermo Scientific Chemicals
Synonyms:
Acetic acid sodium salt
AC21266-0010
AC21266-2500
TS21266-0010EA
103
USD
TS21266-0010
TS21266-2500
Sodium acetate, anhydrous 99% for biochemistry
Sodium acetate
Formula:
H₃CCOONa MW: 82.03 g/mol Boiling Pt: 881.4 Melting Pt: 324 Flash Pt: >250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012459 CAS Number: 127-09-3 Merck Index: 12,08711 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤1 % |
UV | at 280 nm A: ≤0.01 |
Titration with HClO4 | ≥98.5 % |
Heavy metals (as Pb) | ≤0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...