Sodium acetate, anhydrous 99+% for analysis

Supplier: Thermo Scientific Chemicals

Synonyms: Acetic acid sodium salt

AC22089-0010 AC22089-0050 AC22089-2500
TS22089-0010EA 146 USD
TS22089-0010 TS22089-0050 TS22089-2500
Sodium acetate, anhydrous 99+% for analysis
Sodium acetate
Formula: H₃CCOONa
MW: 82.03 g/mol
Boiling Pt: 881.4
Melting Pt: 324
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
EINECS: 204-823-8
REACH: 01-2119485123-42
Merck Index: 12,08711

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤1 %
pH 7.5 to 9.2 (5 % at 20°C)
Titration with HClO4 ≥99.0 %
Chloride (Cl) ≤10 ppm
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤20 ppm
Aluminium (Al) ≤10 ppm
Calcium (Ca) ≤10 ppm
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤5 ppm
Potassium (K) ≤200 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR