Dimethyl sulphide 99+%, extra pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: DMS

AC12776-0010 AC12776-0025 AC12776-0100
TS12776-0010EA 66 USD
TS12776-0010 TS12776-0025 TS12776-0100
Dimethyl sulphide 99+%, extra pure
Dimethyl sulphide
Formula: C₂H₆S
MW: 62.14 g/mol
Boiling Pt: 36.2
Melting Pt: -98
Density: 0.846 g/cm³
Flash Pt: –48 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008562
CAS Number: 75-18-3
EINECS: 200-846-2
UN: 1164
ADR: 3 II
Merck Index: 12,06204

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4341 to 1.4361 (20°C, 589 nm)
Water ≤0.3 %
GC ≤0.2 % Methylmercaptan


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR