Trimethyltin chloride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC16399-0100
AC16399-0500
TS16399-0100EA
86
USD
TS16399-0100
TS16399-0500
Trimethyltin chloride 99%
Trimethyltin chloride
Formula:
C₃H₉ClSn Boiling Pt: 148 °C (760 mmHg) Melting Pt: 37…39 °C Flash Pt: 97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000520 CAS Number: 1066-45-1 EINECS: 213-917-8 UN: 3146 ADR: 6.1,II |
Specification Test Results
Appearance | White Crystals |
Infrared spectrum | Conforms |
Melting point | 37 to 39 °C |
GC | ≥98.5 % |
Solubility | Clear colorless |
TLC | Complies |
Residue after ignition | ≤1.0 % |
pH | 6.5 to 8.5 (1 % in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...