Cerium(IV) sulphate tetrahydrate 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cerium disulphate tetrahydrate, Ceric sulphate tetrahydrate

AC19913-0025 AC19913-1000 AC19913-5000
TS19913-0025EA 3142 USD
TS19913-0025 TS19913-1000 TS19913-5000
Cerium(IV) sulphate tetrahydrate 98+%
Cerium(IV) sulphate tetrahydrate
Formula: Ce(SO₄)₂·4H₂O
MW: 404.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149427
CAS Number: 10294-42-5
EINECS: 237-029-5
UN: 3260
Merck Index: 12,02036

Order Now

Specification Test Results

Appearance Bright yellow Powder
Titration Iodometric ≥98.0 %
Chloride (Cl) ≤100 ppm
Copper (Cu) ≤30 ppm
Iron (Fe) ≤150 ppm
Lead (Pb) ≤30 ppm
Nickel (Ni) ≤30 ppm
Heavy metals (as Pb) ≤30 ppm
Insoluble matter ≤0.1 % (in H2SO4)
Subst not pptd by NH4OH ≤0.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR