Calcium sulfate dihydrate 98+%, Extra Pure
Supplier: Thermo Scientific Chemicals
Synonyms:
Plaster of Paris
AC44638-0010
AC44638-0050
TS44638-0010EA
65
USD
TS44638-0010
TS44638-0050
Calcium sulfate dihydrate 98+%, Extra Pure
Calcium sulphate dihydrate
Formula:
CaSO₄·2H₂O MW: 172.17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00149625 CAS Number: 10101-41-4 |
Specification Test Results
Appearance | White Powder |
Titration Complexometric | ≥98.0 % |
Water | 19 to 23 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...