Calcium bromide hydrate for analysis

Supplier: Thermo Scientific Chemicals
Danger

AC30084-0010 AC30084-0025 AC30084-5000
TS30084-0010EA 137 USD
TS30084-0010 TS30084-0025 TS30084-5000
Calcium bromide hydrate for analysis
Calcium bromide hydrate
Formula: Br₂Ca·H₂O
Melting Pt: 730 °C
Storage Temperature: Ambient
MDL Number: MFCD00149608
CAS Number: 71626-99-8
EINECS: 232-164-6

Order Now

Specification Test Results

Appearance White Crystalline powder and/or chunks
Titration Complexometric ≥83.0 % (CaBr2)
Acidity/alkalinity Passes test
Chloride (Cl) ≤2000 ppm
Sulfate (SO4) ≤100 ppm
Bromate and Chlorate Passes test
Arsenic (As) ≤2 ppm
Barium (Ba) Passes test
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm
Subst not pptd ≤0.4 % By ammonium oxalate (as SO4)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR