Bismuth (III) sulfide 99%, pure
Supplier: Thermo Scientific Chemicals
Synonyms:
Dibismuth trisulphide
AC45399-1000
AC45399-5000
TS45399-1000EA
137
USD
TS45399-1000
TS45399-5000
Bismuth (III) sulfide 99%, pure
Bismuth(III) sulphide
Formula:
Bi₂S₃ MW: 514.16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00014204 CAS Number: 1345-07-9 |
Specification Test Results
Appearance | Gray Powder and/or chunks |
Titration Complexometric | 80.1 to 82.5 % (Bi) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...