Bismuth(III) oxide 98+%, pure
Supplier: Thermo Scientific Chemicals
Synonyms:
Bismuth Trioxide, Dibismuth trioxide
AC19262-0010
AC19262-0050
TS19262-0010EA
141
USD
TS19262-0010
TS19262-0050
Bismuth(III) oxide 98+%, pure
Bismuth(III) oxide
Formula:
BiO₃ MW: 465.96 g/mol Boiling Pt: 1890 Melting Pt: 825 Density: 8.9 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00003462 CAS Number: 1304-76-3 EINECS: 215-134-7 Merck Index: 13,01273 |
Specification Test Results
Appearance | Yellow Powder |
Assay | 98.0 to 102.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...