Bismuth(III) bromide 97%

Supplier: Thermo Scientific Chemicals
Danger

AC44777-0250 AC44777-1000
TS44777-0250EA 63 USD
TS44777-0250 TS44777-1000
Bismuth(III) bromide 97%
Bismuth(III) bromide
Formula: BiBr₃
MW: 448.69 g/mol
Boiling Pt: 278
Melting Pt: 218
Storage Temperature: Ambient
MDL Number: MFCD00014195
CAS Number: 7787-58-8
UN: 3260
ADR: 8 II
Merck Index: 12,01299

Order Now

Specification Test Results

Appearance Light yellow to yellow or light gray Crystalline powder or crystals
Melting point 218 to 220°C
Assay 96 to 103 % (Bi)
Chloride (Cl) ≤0.1 %
Iron (Fe) ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR